Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL4, Rat, Monoclonal Antibody, Abnova™

Catalog No. 89113485 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-485 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-485 Supplier Abnova Corporation Supplier No. MAB8546

Rat monoclonal antibody raised against Il4.

Specifications

Antigen IL4
Applications ELISA, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Description Rat monoclonal antibody raised against Il4.
Dilution ELISA (1:1000-1:2000) Western Blot (2-5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS, pH 7.5, 0.1% BSA
Gene Il4
Gene Alias Il-4
Gene Symbols Il4
Host Species Rat
Immunogen Whole expressing mouse Il4.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16189
Test Specificity This antibody shows no cross-reactivity with human IL-4 and other unrelated proteins.
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.