Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL6, Rat, Clone: MP5-20F3, Abnova™

Catalog No. 89111335 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-335 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-335 Supplier Abnova Corporation Supplier No. MAB3310
Add to Cart
Add to Cart

Rat monoclonal antibody raised against recombinant Il6.

Specifications

Antigen IL6
Applications Flow Cytometry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone MP5-20F3
Conjugate Unconjugated
Description Rat monoclonal antibody raised against recombinant Il6.
Dilution Western Blot (1-5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 100mM BBS, pH 8.2
Gene Il6
Gene Alias Il-6
Gene Symbols Il6
Host Species Rat
Immunogen Recombinant protein corresponding to mouse Il6.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16193
Test Specificity Mouse Interleukin-6.
Target Species Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.