Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin beta 4/CD104 Antibody (439-9B) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody has been used in 3 publications
$499.50
Specifications
| Antigen | Integrin beta 4/CD104 |
|---|---|
| Clone | 439-9B |
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry < = 1 ug/10^6 cells in 100 ul, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP14336901
![]() |
Novus Biologicals
NBP1433690.1MG |
0.1 mg |
Each for $499.50
|
|
|||||
NBP14336903
![]() |
Novus Biologicals
NBP1433690.025MG |
0.025 mg | N/A | N/A | N/A | ||||
Description
Integrin beta 4/CD104 Monoclonal specifically detects Integrin beta 4/CD104 in Human, Bovine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Integrin beta 4/CD104 | |
| 0.5 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| RUO | |
| Human, Bovine | |
| P16144 | |
| 3691 | |
| The immunogen for this antibody was CD104. | |
| Primary | |
| Store at 4C. Do not freeze. |
| 439-9B | |
| Western Blot 1:100-1:2000, Flow Cytometry < = 1 ug/10^6 cells in 100 ul, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Monoclonal | |
| Rat | |
| Adaptive Immunity, Immunology, Signal Transduction | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| CD104, CD104 antigen, GP150, integrin beta-4, integrin beta-4 subunit, integrin, beta 4 | |
| ITGB4 | |
| IgG2b | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title