Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Itga2, Rat, PE, Clone: DX5, Abnova™

Catalog No. 89112453 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-453 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-453 Supplier Abnova Corporation Supplier No. MAB5782
Add to Cart
Add to Cart

Rat monoclonal antibody raised against Itga2.

Specifications

Antigen Itga2
Applications Flow Cytometry
Classification Monoclonal
Clone DX5
Conjugate PE
Description Rat monoclonal antibody raised against Itga2.
Dilution Flow Cytometry (0.2 ug/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Itga2
Gene Alias CD49B/DX5
Gene Symbols Itga2
Host Species Rat
Immunogen RIL-2-propagated NK1.1+ cells from C57BL/6 mice.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16398
Test Specificity Mouse CD49b, Mr 160-KDa. The DX5 antibody is not suitable for immunohistochemical staining of either frozen or paraffin sections.
Target Species Mouse
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.