Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Itga5, Rat, PE, Clone: 5H10-27 (MFR5), Abnova™

Catalog No. 89113836 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-836 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-836 Supplier Abnova Corporation Supplier No. MAB9874
Add to Cart
Add to Cart

Rat monoclonal antibody raised against Itga5.

Specifications

Antigen Itga5
Applications Flow Cytometry
Classification Monoclonal
Clone 5H10-27 (MFR5)
Conjugate PE
Description Rat monoclonal antibody raised against Itga5.
Dilution Flow Cytometry (≤ 0.3 ug/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Itga5
Gene Alias Cd49e/Fnra
Gene Symbols Itga5
Host Species Rat
Immunogen Native Itga5 from mouse mast cell line MC/9.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16402
Target Species Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.