Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Itgal, Rat, Clone: M17/4, Abnova™

Catalog No. 89113235 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-235 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-235 Supplier Abnova Corporation Supplier No. MAB7916
Add to Cart
Add to Cart

Rat monoclonal antibody raised againt native Itgal.

Specifications

Antigen Itgal
Applications Blocking Assay, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone M17/4
Conjugate Unconjugated
Description Rat monoclonal antibody raised againt native Itgal.
Dilution Immunohistochemistry Immunoprecipitation Flow Cytometry (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene Itgal
Gene Alias Cd11a/LFA-1/Ly-15/Ly-21
Gene Symbols Itgal
Host Species Rat
Immunogen Native purified Itgal from C57BL/6 mouse splenic secondary cytotoxic T lymphocytes.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16408
Test Specificity This antibody reacts with Itgal (alpha-subunit of murine LFA-1), a180 KDa type I transmembrane glycoprotein expressed on B and T lymphocytes, monocytes, macrophages, neutrophils, basophils and eosinophils.
Target Species Mouse
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.