Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Itgam, Rat, Clone: M1/70, Abnova™

Catalog No. 89111272 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-272 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-272 Supplier Abnova Corporation Supplier No. MAB3224
Add to Cart
Add to Cart

Rat monoclonal antibody raised against Itgam.

Specifications

Antigen Itgam
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone M1/70
Conjugate Unconjugated
Description Rat monoclonal antibody raised against Itgam.
Dilution Flow Cytometry (1 ug/106 cells) The optimal working dilution should be determined by the end user.
Formulation In 100mM BBS, pH 8.2
Gene Itgam
Gene Alias CD11b/CD18/CR3/CR3A/Cd11b/F730045J24Rik/Ly-40/MAC1/Mac-1/Mac-1a
Gene Symbols Itgam
Host Species Rat
Immunogen Native Itgam from mouse splenic T cells and splenocytes.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16409
Test Specificity alphaM subunit (Mr 170 KDa) of the CD11b/CD18 heterodimer.
Target Species Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.