Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Itgb1, Rat, Clone: KMI6, Abnova™

Catalog No. 89109164 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-164 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-164 Supplier Abnova Corporation Supplier No. MAB10950

Rat monoclonal antibody raised against Itgb1.

Specifications

Antigen Itgb1
Applications Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone KMI6
Conjugate Unconjugated
Description Rat monoclonal antibody raised against Itgb1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In aqueous buffer (0.09% sodium azide)
Gene Itgb1
Gene Alias 4633401G24Rik/AA409975/AA960159/CD29/Fnrb/gpIIa
Gene Symbols Itgb1
Host Species Rat
Immunogen Mouse Itgb1.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16412
Test Specificity The monoclonal antibody reacts with mouse Itgb1, a 120-130 kDa member of the beta integrin family expressed by leukocytes, endothelial, smooth muscle and epithelial cells.
Target Species Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.