Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Itgb2, Rat, FITC, Clone: C71/16, Abnova™

Catalog No. 89113831 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-831 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-831 Supplier Abnova Corporation Supplier No. MAB9869
Add to Cart
Add to Cart

Rat monoclonal antibody raised against native Itgb2.

Specifications

Antigen Itgb2
Applications Flow Cytometry
Classification Monoclonal
Clone C71/16
Conjugate FITC
Description Rat monoclonal antibody raised against native Itgb2.
Dilution Flow Cytometry (≤ 1 ug/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Itgb2
Gene Alias 2E6/AI528527/Cd18/LAD/LCAMB/Lfa1/MF17
Gene Symbols Itgb2
Host Species Rat
Immunogen Native Itgb2 from cell membrane of mouse T cell lymphoma BW5147.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16414
Target Species Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.