Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kdr, Rat, Clone: Avas12a1, Abnova™

Catalog No. 89109165 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-165 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-165 Supplier Abnova Corporation Supplier No. MAB10951

Rat monoclonal antibody raised against Kdr.

Specifications

Antigen Kdr
Applications Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone Avas12a1
Conjugate Unconjugated
Description Rat monoclonal antibody raised against Kdr.
Dilution The optimal working dilution should be determined by the end user.
Formulation In aqueous buffer (0.09% sodium azide)
Gene Kdr
Gene Alias 6130401C07/Flk-1/Flk1/Krd-1/Ly73/VEGFR-2/VEGFR2
Gene Symbols Kdr
Host Species Rat
Immunogen Mouse Kdr.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16542
Test Specificity The monoclonal antibody reacts with mouse Kdr.
Target Species Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.