Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Klrc1/Klrc2/Klrc3, Rat, Biotin, Clone: 20d5, Abnova™

Catalog No. 89112444 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-444 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-444 Supplier Abnova Corporation Supplier No. MAB5773
Add to Cart
Add to Cart

Rat monoclonal antibody raised against Klrc1/Klrc2/Klrc3.

Specifications

Antigen Klrc1/Klrc2/Klrc3
Applications Flow Cytometry
Classification Monoclonal
Clone 20d5
Conjugate Biotin
Description Rat monoclonal antibody raised against Klrc1/Klrc2/Klrc3.
Dilution Flow Cytometry (1 ug/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Alias CD159a/NKG2A/NKG2B
Gene Symbols Klrc1
Host Species Rat
Immunogen CHO cells transfected with murine NKG2.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16641/16642/58179
Test Specificity Mouse NKG2 (Mr ∼ 43 KDa).
Target Species Mouse
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.