Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rat anti-LAG3 (Azide Free), Clone: C9B7w,

Catalog No. MABF846MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF846MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF846MI Supplier MilliporeSigma™ Supplier No. MABF846
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Specifically detects LAG3 (Azide Free) clone: C9B7w in Mouse samples, and it is validated for Activity Assay, Flow Cytometry

Lymphocyte-activation gene 3 (LAG3), also known as cluster of differentiation 223 (CD223), is an immunoglobulin superfamily (IgSF) type I membrane protein encoded by the LAG3 gene in human. LAG3 is expressed in human activated T and NK cells and involved lymphocyte activation by functioning as a ligand for major histocompatibility complex (MHC) class II molecules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAG3 (Azide Free)
Applications Functional Assay, Flow Cytometry
Classification Monoclonal
Clone C9B7w
Formulation Purified rat monoclonal IgG1 in buffer containing PBS without preservatives.
Gene Accession No. Q61790
Gene Symbols Lag3
Host Species Rat
Immunogen Recombinant protein corresponding to within the extracellular D2 domain of mouse LAG3.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_032505
Target Species Mouse
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.