Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lamp1, Rat, FITC, Clone: 1D4B, Abnova™

Catalog No. 89112478 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-478 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-478 Supplier Abnova Corporation Supplier No. MAB5835
Add to Cart
Add to Cart

Rat monoclonal antibody raised against native Lamp1.

Specifications

Antigen Lamp1
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone 1D4B
Conjugate FITC
Description Rat monoclonal antibody raised against native Lamp1.
Dilution Flow Cytometry (1 ug/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Lamp1
Gene Alias AI196048/CD107a/Lamp-1
Gene Symbols Lamp1
Host Species Rat
Immunogen Native purified Lamp1 from plasma membrane fraction of mouse embryo NIH3T3 cell line.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16783
Test Specificity Murine CD107a/LAMP-1 (Mr 110-140 KDa with a core protein of Mr 40 KDa).
Target Species Mouse
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.