Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lgi4, Rat, Clone: KT18, Abnova™

Catalog No. 89110563 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-563 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-563 Supplier Abnova Corporation Supplier No. MAB1667
Add to Cart
Add to Cart

Rat monoclonal antibody raised against synthetic peptide of Lgi4.

Specifications

Antigen Lgi4
Applications Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone KT18
Conjugate Unconjugated
Description Rat monoclonal antibody raised against synthetic peptide of Lgi4.
Dilution Western Blot (1:500) Immunohistochemistry (Frozen section) (1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Lgi4
Gene Alias Lgil3/MGC107385/clp
Gene Symbols Lgi4
Host Species Rat
Immunogen A synthetic peptide (conjugated with BSA) corresponding to mouse Lgi4.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 243914
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.