Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LTBR, Rat, Clone: Carlos-1, Abnova™

Catalog No. 89111561 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-561 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-561 Supplier Abnova Corporation Supplier No. MAB3721

Rat monoclonal antibody raised against recombinant LTBR.

The protein encoded by this gene is a member of the tumor necrosis factor (TNF) family of receptors. It is expressed on the surface of most cell types, including cells of epithelial and myeloid lineages, but not on T and B lymphocytes. The protein specifically binds the lymphotoxin membrane form (a complex of lymphotoxin-alpha and lymphtoxin-beta). The encoded protein and its ligand play a role in the development and organization of lymphoid tissue and tranformed cells. Activation of the encoded protein can trigger apoptosis. [provided by RefSeq

Specifications

Antigen LTBR
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone Carlos-1
Conjugate Unconjugated
Description Rat monoclonal antibody raised against recombinant LTBR.
Dilution Flow Cytometry (1:1000) Immunoprecipitation (1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene LTBR
Gene Alias D12S370/LT-BETA-R/TNF-R-III/TNFCR/TNFR-RP/TNFR2-RP/TNFRSF3
Gene Symbols LTBR
Host Species Rat
Immunogen Recombinant human IgG1 Fc fusion protein corresponding to amino acids 28-220 of human LTBR.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4055
Test Specificity Recognizes human LTbetaR
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.