Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly6c1, Rat, Clone: HK1.4, Abnova™

Catalog No. 89112735 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-735 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-735 Supplier Abnova Corporation Supplier No. MAB6207
Add to Cart
Add to Cart

Rat monoclonal antibody raised against native Ly6c1.

Specifications

Antigen Ly6c1
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone HK1.4
Conjugate Unconjugated
Description Rat monoclonal antibody raised against native Ly6c1.
Dilution Flow Cytometry (1 ug/106 cells) The optimal working dilution should be determined by the end user.
Formulation In 100mM BBS, pH 8.2
Gene Ly6c1
Gene Alias AA682074/AA959465/Ly-6C/Ly6c
Gene Symbols Ly6c1
Host Species Rat
Immunogen Native purified Ly6c1 from L3 cells.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17067
Test Specificity Murine Ly-6C, Mr 18 KDa.
Target Species Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2c
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.