Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly6c1/Ly6c2, Rat, Clone: ER-MP20, Abnova™

Catalog No. 89112157 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-157 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-157 Supplier Abnova Corporation Supplier No. MAB5179
Add to Cart
Add to Cart

Rat monoclonal antibody raised against native Ly6c1/Ly6c2.

Specifications

Antigen Ly6c1/Ly6c2
Applications ELISA, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone ER-MP20
Conjugate Unconjugated
Description Rat monoclonal antibody raised against native Ly6c1/Ly6c2.
Dilution Recommend dilution at 1:50 The optimal working dilution should be determined by the end user.
Formulation In PBS (0.1% BSA, 0.02% sodium azide)
Gene Alias AA682074/AA959465/Ly-6C/Ly6c
Gene Symbols Ly6c1
Host Species Rat
Immunogen Native purified Ly6c1/Ly6c2 from mouse macrophage cell lines.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17067/100041546
Test Specificity The monoclonal antibody ER-MP20 specifically reacts with mouse macrophage precursor cells in the mid-stage of their development (late CFU-M, monoblasts and monocytes). The antigen is a 14 kD surface protein which is very similar to Ly-6C and may be analogous to human CD59. The monoclonal antibody ER-MP20 has been raised after immunization of rats with mouse macrophage cell lines and reacts with mouse macrophage precursor cells. The monoclonal antibody also identifies activated macrophages in inflammatory tissues where the simultaneous use of the murine pan-macrophage marker BM8 (anti-F4/80) is recommended.
Target Species Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.