Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly76, Rat, Clone: TER-119, Abnova™

Catalog No. 89109222 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-222 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-222 Supplier Abnova Corporation Supplier No. MAB11007

Rat monoclonal antibody raised against Ly76.

Specifications

Antigen Ly76
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone TER-119
Conjugate Unconjugated
Description Rat monoclonal antibody raised against Ly76.
Dilution The optimal working dilution should be determined by the end user.
Formulation In aqueous buffer (0.09% sodium azide)
Gene Ly76
Gene Alias TER-119/Ter119
Gene Symbols Ly76
Host Species Rat
Immunogen Mouse Ly76.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104231
Target Species Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.