Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPRE3, Rat, Clone: KT36, Abnova™

Catalog No. 89110571 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-571 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-571 Supplier Abnova Corporation Supplier No. MAB1677
Add to Cart
Add to Cart

Rat monoclonal antibody raised against full-length recombinant MAPRE3.

The protein encoded by this gene is a member of the RP/EB family of genes. The protein localizes to the cytoplasmic microtubule network and binds APCL, a homolog of the adenomatous polyposis coli tumor suppressor gene. [provided by RefSeq

Specifications

Antigen MAPRE3
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone KT36
Conjugate Unconjugated
Description Rat monoclonal antibody raised against full length recombinant MAPRE3.
Dilution Immunofluorescence (1:100-1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene MAPRE3
Gene Alias EB3/EBF3/EBF3-S/RP3
Gene Symbols MAPRE3
Host Species Rat
Immunogen Recombinant GST fusion protein corresponding to full length human MAPRE3.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22924
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.