Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Marcks, Rat, Clone: MAR 11/2, Abnova™

Catalog No. 89110587 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-587 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-587 Supplier Abnova Corporation Supplier No. MAB1713
Add to Cart
Add to Cart

Rat monoclonal antibody raised against synthetic peptide of Marcks.

Specifications

Antigen Marcks
Applications Western Blot
Classification Monoclonal
Clone MAR 11/2
Conjugate Unconjugated
Description Rat monoclonal antibody raised against synthetic peptide of Marcks.
Dilution Immunohistochemistry (1-5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.1% BSA, 0.1% proclin)
Gene Marcks
Gene Alias KINC/Macs
Gene Symbols Marcks
Host Species Rat
Immunogen A synthetic peptide corresponding to rat Marcks.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25603
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.