Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MHC Class II (I-A/I-E), Rat, Clone: M5/114.15.2, Abnova™

Catalog No. 89134831 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-831 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-831 Supplier Abnova Corporation Supplier No. MAB11076

Rat monoclonal antibody raised against MHC Class II (I-A/I-E).

Specifications

Antigen MHC Class II (I-A/I-E)
Applications Flow Cytometry, Functional Assay, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone M5/114.15.2
Conjugate Unconjugated
Description Rat monoclonal antibody raised against MHC Class II (I-A/I-E).
Dilution The optimal working dilution should be determined by the end user.
Formulation In aqueous buffer (0.09% sodium azide)
Host Species Rat
Immunogen Mouse MHC Class II (I-A/I-E).
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.