Learn More
CD102 Rat anti-Mouse, Brilliant Violet 421, Clone: 3C4(mIC2/4), BD Optibuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 740018
Description
The 3C4 (mIC2/4) monoclonal antibody specifically binds to the mouse ICAM-2 (CD102) cell surface glycoprotein, a ligand for LFA-1. CD102 is constitutively expressed on endothelial cells, T and B lymphocytes, and alveolar walls. It is also expressed on a variety of leukocyte cell lines. CD102 does not appear to be involved in the development of hematopoietic cells. In a model for allergic asthma, endothelial CD102 mediates the transmigration of eosinophils (but not lymphocytes, monocytes, or macrophages) into the airway lumen. The 3C4 (mIC2/4) antibody blocks interactions between ICAM-2 and LFA-1.
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.
Specifications
| CD102 | |
| Monoclonal | |
| 0.2mg/mL | |
| ICAM-2 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| 3C4(mIC2/4) | |
| Brilliant Violet 421 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Transfected Cell Line | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only