Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD102 Rat anti-Mouse, Brilliant Violet 510, Clone: 3C4(mIC2/4), BD Optibuild™

Catalog No. BDB740116 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740116 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740116 Supplier BD Biosciences Supplier No. 740116
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 3C4 (mIC2/4) monoclonal antibody specifically binds to the mouse ICAM-2 (CD102) cell surface glycoprotein, a ligand for LFA-1. CD102 is constitutively expressed on endothelial cells, T and B lymphocytes, and alveolar walls. It is also expressed on a variety of leukocyte cell lines. CD102 does not appear to be involved in the development of hematopoietic cells. In a model for allergic asthma, endothelial CD102 mediates the transmigration of eosinophils (but not lymphocytes, monocytes, or macrophages) into the airway lumen. The 3C4 (mIC2/4) antibody blocks interactions between ICAM-2 and LFA-1.

The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405-nm and Em Max at 510-nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50-nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD102
Applications Flow Cytometry
Classification Monoclonal
Clone 3C4(mIC2/4)
Concentration 0.2mg/mL
Conjugate Brilliant Violet 510
Description ICAM-2
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.