Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD103 Rat anti-Mouse, Brilliant Violet 605, Clone: M290, BD Optibuild™

Catalog No. BDB740355 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740355 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740355 Supplier BD Biosciences Supplier No. 740355
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The M290 antibody specifically binds to CD103, the α chain of α IEL β 7 integrin. CD103 has a unique and fairly restricted tissue distribution. It is expressed on almost all intestinal intraepithelial lymphocytes (IEL), dendritic epidermal T cells (DEC), subpopulations of peripheral T cells, and distinct subsets of fetal, neonatal, and adult thymocytes. E-cadherin is the epithelial cell ligand for α IEL β 7 integrin. The ordered expression of α IEL during thymocyte development (which occurs under the influence of the thymic epithelium), high level of α α IEL β 7-mediated adhesion in in vitro assays.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD103
Applications Flow Cytometry
Classification Monoclonal
Clone M290
Concentration 0.2mg/mL
Conjugate Brilliant Violet 605
Description Itgae; Integrin alpha-E; αE; alpha-E1; ITAE; Integrin αIEL chain; aM290
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse Intestinal Epithelial Cells
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.