Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11a Rat anti-Mouse, Brilliant Violet 711, Clone: M17/4, BD Optibuild™

Catalog No. BDB740676 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740676 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740676 Supplier BD Biosciences Supplier No. 740676
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The M17/4 antibody reacts with the 180 kDa α L chain of LFA-1 (CD11a/CD18, α L β 2 integrin), a heterodimeric surface glycoprotein expressed on almost all leukocytes. CD8a+CD8b- intestinal intraepithelial T lymphocytes, which are believed to be thymus independent, do not express CD11a. LFA-1 mediates a variety of heterotypic and homotypic intercellular adhesions through interaction with ICAM-1 (CD54) and ICAM-2 (CD102), including participation in the immunological synapses between CD8+ T lymphocytes and antigen-presenting cells. mAb M17/4 blocks a variety of LFA-1-mediated cells interactions in vitro, and costimulatory effects have also been described. In vivo treatment with M17/4 mAb reduces the severity of graft-versus-host reactions, prolongs allograft survival, inhibits the development of autoimmunity, and blocks substance P-induced leukocyte migration. The M17/4 and 2D7 (Cat. No. 553120) antibodies are reported to recognize different epitopes of the CD11a molecule.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11a
Applications Flow Cytometry
Classification Monoclonal
Clone M17/4
Concentration 0.2mg/mL
Conjugate Brilliant Violet 711
Description Itgal; Integrin alpha-L; Integrin αL; ITAL; LFA-1A; LFA-1α; Ly-15
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen C57BL/6 Mouse Splenic Secondary Cytotoxic T Lymphocytes
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.