Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD125 Rat anti-Mouse, Brilliant Violet 786, Clone: T21, BD Optibuild™

Catalog No. BDB741025 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB741025 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB741025 Supplier BD Biosciences Supplier No. 741025
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The T21 monoclonal antibody specifically binds to CD125 which is also known as the alpha subunit of the IL-5 Receptor (IL5RA/IL-5R α ). CD125 is a type I transmembrane protein that belongs to the hemopoietin receptor and Ig gene superfamilies. It is expressed on B cells and eosinophils. CD125 associates with the Common β chain ( β c/Bc) subunit, also known as CD131, to form the functional IL-5 Receptor complex. Mouse IL-5 belongs to the hematopoietic growth factor family of cytokines and promotes the growth and differentiation of B cells and eosinophils. Upon binding, the T21 antibody blocks IL-5 binding to its receptor and can inhibit IL-5 induced B-cell proliferation and antibody production.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD125
Applications Flow Cytometry
Classification Monoclonal
Clone T21
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Description Il5r; Il5ra; IL-5RA; IL-5Rα; IL-5R-alpha; IL-5R subunit alpha
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse T88-M Cell Line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 λ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.