Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD126 Rat anti-Mouse, Brilliant Violet 786, Clone: D7715A7, BD Optibuild™

Catalog No. BDB740910 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740910 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740910 Supplier BD Biosciences Supplier No. 740910
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The D7715A7 monoclonal antibody specifically binds to the mouse IL-6 receptor, which is also known as, CD126. The immunogen used to generate the D7715A7 hybridoma was OKT4 hybridoma cells. The D7715A7 hybridoma was selected based on its capacity to produce antibody that inhibited IL-6 binding to 2F4 cells (mouse B-cell hybridoma expressing high levels of IL-6 receptors). The binding of purified D7715A7 to B9 cells (mouse B-cell hybridoma expressing high levels of IL-6 receptors) is inhibited by recombinant mouse IL-6. This antibody has been reported to inhibit the in vitro and in vivo growth of the IL-6-dependent plasmacytoma line, T1033C2.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD126
Applications Flow Cytometry
Classification Monoclonal
Clone D7715A7
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Description Il6ra; IL-6 Receptor α chain; IL-6RA; IL-6R alpha
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen OKT4 hybridoma cells
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.