Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD13 Rat anti-Mouse, Brilliant Violet 605, Clone: R3-242, BD Optibuild™

Catalog No. BDB740359 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740359 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740359 Supplier BD Biosciences Supplier No. 740359
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The R3-242 monoclonal antibody specifically binds to aminopeptidase N (APN), which has been reported to be the mouse CD13 homologue. APN is expressed in a variety of mammalian tissues, including intestinal and renal epithelial brush border, endothelium, liver, lung, and brain. In lymphoid tissues, R3-242 mAb is reported to detect APN on monocytes, macrophages, mast cells, dendritic cells, and B lymphocytes, but not on T cells, and thymocytes. However, the reported reactivity with B cells may be non-specific, via binding to the Fc γreceptors (CD16/CD32). This observation is in agreement with reports using different antibodies to mouse APN . APN may play a role in antigen presentation in the immune system. R3-242 mAb does not inhibit the enzymatic activity of APN.

This antibody is conjugated to BD Horizon™ BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max of 602-nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20-nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605-nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD13
Applications Flow Cytometry
Classification Monoclonal
Clone R3-242
Concentration 0.2mg/mL
Conjugate Brilliant Violet 605
Description Anpep; Alanyl aminopeptidase; Aminopeptidase N; AP-N; mAPN; Lap-1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Crude Aminopeptidase N Preparation from BALB/c Mouse Intestinal Epithelium
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.