Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD132 Rat anti-Mouse, Brilliant Violet 421, Clone: TUGm2, BD Optibuild™

Catalog No. BDB740039 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740039 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740039 Supplier BD Biosciences Supplier No. 740039
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The TUGm2 monoclonal antibody specifically binds to CD132, which is also known as, the common γsubunit ( γc). The γc chain subunit is shared by the IL-2, IL-4, IL-7, IL-9, and IL-15 receptor complexes. The γc receptor is expressed constitutively at low levels by most lymphocytes, myeloid cells, embryonic thymocytes, and lymphoid cell lines. The γc receptor is a 75-80 kD transmembrane glycoprotein which mediates signal transducing activities of cytokine receptor complexes with which it is associated. TUGm2 antibody can significantly block the binding of and proliferative response to IL-4 by CTLL-2 cells. It can also inhibit binding to high-affinity IL-2 receptors expressed by CTLL-2 cells. By itself, the TUGm2 antibody reportedly fails to significantly inhibit CTLL-2 proliferative responses to IL-2. However, when added along with TM- β 1 (Cat. No. 553359), specific for the mouse IL-2R β chain, TUGm2 was shown to synergistically reduce IL-2-driven CTLL-2 proliferative responses. TUGm2 can reduce the IL-7-induced proliferation of Con A-activated mouse splenocytes and the IL-7 responsive pre-B cell line, IxN/2b.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD132
Applications Flow Cytometry
Classification Monoclonal
Clone TUGm2
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Description Il2rg; IL-2Rγ; Commin gamma chain; Common γ; gamma c receptor; γc
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse IL-2Rγ Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.