Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD134 Rat anti-Mouse, Brilliant Violet 650, Clone: OX-86, BD Optibuild™

Catalog No. BDB740545 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740545 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740545 Supplier BD Biosciences Supplier No. 740545
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The OX-86 monoclonal antibody specifically binds to the OX-40 antigen (CD134), also known as OX-40L receptor. CD134 is a 50-kDa type-I membrane glycoprotein that belongs to the NGFR/TNFR superfamily. Mouse CD134 is expressed on activated CD4+ and CD8+ T lymphocytes and has been shown to be the sole receptor for the OX-40 Ligand (OX-40L). In the brains of mice with actively induced experimental allergic encephalomyelitis, the expression of CD134 on CD4+ T lymphocytes correlates with disease progression. The OX-40/OX-40L system supplies a costimulatory signal for T-cell proliferation and B-cell proliferation and differentiation. In addition, OX-40 antigen provides a costimulatory signal that induces T cells to proliferate in a CD28-independent manner. In the intact animal, CD134 does not appear to be essential for many T-cell responses, but it seems to play a major role in the pathogenesis of some autoimmune diseases. The OX-86 mAb stains both CD4+ and CD8+ activated T cells, and this expression pattern has been confirmed using OX-40L-Ig fusion protein. CD134 was also detected, using OX-86 mAb, on B cells after stimulation with anti-IgM plus anti-CD40 mAb HM40-3 (Cat. no. 553721). OX-86 mAb does not block binding of OX-40L to OX-40, and it stimulates T-cell proliferation mildly.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 650-nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD134
Applications Flow Cytometry
Classification Monoclonal
Clone OX-86
Concentration 0.2mg/mL
Conjugate Brilliant Violet 650
Description Tnfrsf4; Ox40; OX40L receptor; Ly-70; ACT35; Txgp1; TXGP1L
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Recombinant Mouse OX-40 3/4 CD4 Chimeric Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.