Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD137 Rat anti-Mouse, Brilliant Violet 605, Clone: 1AH2, BD Optibuild™

Catalog No. BDB740364 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740364 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740364 Supplier BD Biosciences Supplier No. 740364
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 1AH2 monoclonal antibody specifically binds to CD137. CD137 is a member of the TNFR/NGFR superfamily that is likewise known as Tnfrsf9, 4-1BB, Ly-63, or ILA. Monomers or multimeric forms of CD137 are expressed, upon activation, on the surface of splenic T lymphocytes, thymocytes, intestinal intraepithelial T lymphocytes (IEL), and some T cell lines and clones. While stimulating T cells by IL-2, IL-4, or anti-CD28 alone does not result in the expression of CD137; addition of IL-2, IL-4, anti-CD28, or syngeneic accessory cells to splenic T cells stimulated via TCR/CD3 can result in a high level CD137 expression. CD137 is also reportedly expressed on IL-2 activated NK cells, but not on freshly isolated NK cells. CD137 physically associates with p56 [lck] through a Cys-Arg-Cys-Pro binding site in its cytoplasmic domain; the same motif in the cytoplasmic tail of the CD4 and CD8a molecules is responsible for association with p56 [lck]. A signaling function for the CD137 molecule in mouse T cells is indicated by reports in which cross-linking of CD137 with 1AH2 mAb resulted in enhanced proliferation of CD3e-activated splenic T cells and IEL and in enhanced cytolytic activity of IEL in response to immobilized anti-CD3e. In addition to extracellular matrix proteins which bind to CD137, the CD137L (4-1BBL) serves as a ligand for CD137. This molecule has also been detected on LPS-activated macrophages, and anti-IgM antibody-activated splenic B cells. Interaction between T and B cells through CD137/CD137L is reported to play a role in antigen presentation, further supporting a costimulatory role for CD137 in the immune response of T lymphocytes.

This antibody is conjugated to BD Horizon™ BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max of 602-nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20-nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605-nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD137
Applications Flow Cytometry
Classification Monoclonal
Clone 1AH2
Concentration 0.2mg/mL
Conjugate Brilliant Violet 605
Description 4-1BB; Tnfrsf9; Ly-63; ILA
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Recombinant mouse 4-1BB
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.