Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD140a Rat anti-Mouse, Brilliant Violet 786, Clone: APA5, BD Optibuild™

Catalog No. BDB740930 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740930 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740930 Supplier BD Biosciences Supplier No. 740930
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The APA5 antibody monoclonal antibody specifically binds to CD140a, the Platelet-Derived Growth Factor Receptor alpha chain (PDGFR-a, PDGFR- α ). CD140a is a receptor tyrosine kinase that is widely expressed on cells of mesenchymal origin in the embryo and adult. CD140a is expressed on several other cell types during embryonic development, but not on hematopoietic cells. CD140a binds to PDGF A and B chains, in contrast to PDGFR-b (CD140b) that binds only to the PDGF B chain. Biologically active PDGF is a disulphide-linked dimer, forming the AA, AB, and BB isoforms. Ligand binding to the PDGF Receptor induces the formation of receptor dimers (aa, ab, or bb), autophosphorylation, and internalization. The APA5 antibody has been demonstrated to block binding of PDGF-AA to PDGFR-a-expressing cells in vitro and to block some PDGF-mediated developmental events in vivo.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 786-nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60-nm filter).

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD140a
Applications Flow Cytometry
Classification Monoclonal
Clone APA5
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Description Pdgfra; Pdgfr2; PDGF-R alpha; Platelet derived growth factor receptor alpha
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse PDGF Receptor α chain
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.