Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD146 Rat anti-Mouse, Brilliant Violet 510, Clone: ME-9F1, BD Optibuild™

Catalog No. BDB740200 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740200 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740200 Supplier BD Biosciences Supplier No. 740200
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The ME-9F1 monoclonal antibody specifically binds to mouse CD146. The CD146 adhesion molecule is a type 1 transmembrane glycoprotein and member of the immunoglobulin superfamily. CD146 is expressed by blood vessel endothelial cells and may play roles in forming intercellular junctions between endothelial cells and influencing the transendothelial migration of other cell types. CD146 is also expressed by some melanoma cell lines, NK cells and neutrophils. CD146 is not detectable on mouse monocytes, dendritic cells, T cells, NKT cells, B cells and smooth muscle cells. Increased expression of CD146 is reportedly associated with NK cell maturation and may be used to characterize different functional NK cell subsets. Activated CD146-positive mouse NK cells reportedly are less cytotoxic and secrete less IFN- γthan their CD146-negative counterparts.

The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405-nm and Em Max at 510-nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50-nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD146
Applications Flow Cytometry
Classification Monoclonal
Clone ME-9F1
Concentration 0.2mg/mL
Conjugate Brilliant Violet 510
Description S-endo 1; S-endo; Muc18; Mcam; Gicerin
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen CBA/J Mouse lymph node Cell Line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.