Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD147 Rat anti-Mouse, Brilliant Violet 650, Clone: RL73 (also known as RL73.2), BD Optibuild™

Catalog No. BDB740617 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740617 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740617 Supplier BD Biosciences Supplier No. 740617
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The RL73 monoclonal antibody specifically binds to mouse CD147. CD147 is type I membrane glycoprotein and member of the Ig superfamily. CD147 is also known as extracellular matrix metalloproteinase inducer (EMMPRIN) and gp42/basigin (BSG). The CD147 molecule is widely expressed on a variety of hemopoietic and non-hemopoietic cell types including thymocytes, lymphocytes, monocytes, granulocytes, erythroblasts and erythrocytes, endothelial cells and neoplasms. CD147 reportedly plays significant roles in the reproductive, nervous and immune systems and in tumor progression. In the immune system, CD147 can function as an adhesion molecule and signaling receptor that regulates leukocyte trafficking and immune and inflammatory responses.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 650-nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD147
Applications Flow Cytometry
Classification Monoclonal
Clone RL73 (also known as RL73.2)
Concentration 0.2mg/mL
Conjugate Brilliant Violet 650
Description Bsg; Basigin; BASI; EMMPRIN; gp 42; HT7; HT-7; Neurothelin
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse EL4 Cell Subline
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.