Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD153 Rat anti-Mouse, Brilliant Violet 421, Clone: RM153, BD Optibuild™

Catalog No. BDB740059 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740059 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740059 Supplier BD Biosciences Supplier No. 740059
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The RM153 monoclonal antibody specifically recognizes a 40-kDa type-II membrane glycoprotein, CD30 ligand (CD30L or CD153), which belongs to the NGF/TNF superfamily. In the presence of cytokines, CD153 stimulates B-cell proliferation, antigen-specific antibody production, and polyclonal immunoglobulin secretion. In addition, it costimulates the proliferation of activated T cells. This molecule is expressed on activated T cells, predominantly CD4+, on activated EL4 cells, and on activated Th1 and Th2 cells. The RM153 mAb inhibits the binding of CD153 to CD30.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD153
Applications Flow Cytometry
Classification Monoclonal
Clone RM153
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Description CD30 Ligand; Cd30l; CD30LG; Tnfsf8
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen CHO cells transfected with mouse CD153 gene (Tnfsf8)
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.