Learn More
CD180 Rat anti-Mouse, Brilliant Violet 421, Clone: RP/14, BD Optibuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 740070
Description
The RP/14 monoclonal antibody specifically binds to CD180, the “radioprotective” 105 kDa (RP105) antigen expressed on mature B lymphocytes. Like other members of the Toll-like Receptor (TLR) family, the extracellular region of CD180 contains tandem repeats of a leucine-rich motif flanked by regions containing conserved cysteines. These characteristic structural components of TLR may mediate protein-protein interactions and regulation of signal transduction involved in innate immune responses. Furthermore, CD180 associates with the secretory protein MD-1 on the cell surface. Cross-linking of cell-surface CD180 with the RP/14 antibody induces activation signals in B lymphocytes. Despite the expression of CD180 on lymphocytes of all strains tested (BALB/c, BALB/c. xid , C3H, C57BL/6, CBA/N, and 129/Sv), some strains (BALB/c. xid , C57BL/6. xid , CBA/N, 129/Sv, and 129/Ola) display deficient responses to that stimulatory effects of mAb RP/14.
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.

Specifications
CD180 | |
Monoclonal | |
0.2mg/mL | |
Ly78; Ly-78; Lymphocyte antigen 78; RP105; Radioprotective 105 kDa protein | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
RP/14 | |
Brilliant Violet 421 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
BALB/c mouse splenocytes | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only