Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD180 Rat anti-Mouse, Brilliant Violet 711, Clone: RP/14, BD Optibuild™

Catalog No. BDB740765 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740765 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740765 Supplier BD Biosciences Supplier No. 740765
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The RP/14 monoclonal antibody specifically binds to CD180, the “radioprotective” 105 kDa (RP105) antigen expressed on mature B lymphocytes. Like other members of the Toll-like Receptor (TLR) family, the extracellular region of CD180 contains tandem repeats of a leucine-rich motif flanked by regions containing conserved cysteines. These characteristic structural components of TLR may mediate protein-protein interactions and regulation of signal transduction involved in innate immune responses. Furthermore, CD180 associates with the secretory protein MD-1 on the cell surface. Cross-linking of cell-surface CD180 with the RP/14 antibody induces activation signals in B lymphocytes. Despite the expression of CD180 on lymphocytes of all strains tested (BALB/c, BALB/c. xid , C3H, C57BL/6, CBA/N, and 129/Sv), some strains (BALB/c. xid , C57BL/6. xid , CBA/N, 129/Sv, and 129/Ola) display deficient responses to that stimulatory effects of mAb RP/14.

The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 711-nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD180
Applications Flow Cytometry
Classification Monoclonal
Clone RP/14
Concentration 0.2mg/mL
Conjugate Brilliant Violet 711
Description Ly78; Ly-78; Lymphocyte antigen 78; RP105; Radioprotective 105 kDa protein
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen BALB/c mouse splenocytes
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.