Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD193 Rat anti-Mouse, Brilliant Violet 421, Clone: 83103, BD Optibuild™

Catalog No. BDB740003 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740003 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740003 Supplier BD Biosciences Supplier No. 740003
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 83103 monoclonal antibody specifically binds to CD193, which is also known as, C-C chemokine receptor type 3 (CCR3). CD193 is a chemokine receptor for CCL11/eotaxin, CCL24/eotaxin-2, CCL26/eotaxin-3, CCL5/RANTES, CCL7/MCP-3, CCL8/MCP-2, CCL14/MCP-4, CCL15/HCC-2 and CCL3/MIP-1a. CD193 is reportedly expressed on eosinophils, mast cells and Th2 cells. It plays an important role in eosinophil recruitment to the skin and the lung and in the development of airway hyperresponsiveness.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD193
Applications Flow Cytometry
Classification Monoclonal
Clone 83103
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Description CCR3; CCR-3; chemokine (C-C) receptor 3; CC-CKR3; C-C CKR-3; CKR3; CC-CKR-3
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse CCR3 transfected Cells Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.