Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD1d Rat anti-Mouse, Brilliant Violet 711, Clone: 1B1, BD Optibuild™

Catalog No. BDB740711 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740711 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740711 Supplier BD Biosciences Supplier No. 740711
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 1B1 monoclonal antibody specifically binds to mouse CD1d, a 48-kDa glycoprotein with structural homology to major histocompatibility complex (MHC) class I molecules. The structure, expression, and functions of CD1 antigens are complex and have been reviewed. MAb 1B1 detects CD1d at varying levels on most types of bone marrow and peripheral leukocytes and on epithelial, dendritic, and lymphoid cells in the thymus. It appears to recognize CD1d only in association with β 2m. CD1d has been reported to be expressed by gastrointestinal tract epithelium and in the cytoplasm of hepatocytes via immunohistochemical staining of frozen sections with mAb 3C11 (Cat. No. 559871, for the purified antibody), suggesting a possible role for CD1d in mucosal immunity. However, CD1d expression was not detectable via flow cytometry on intestinal epithelial cells in studies using the anti-CD1d mAbs 3C11, 1B1, and 9C7. The 1B1 antibody competes with mAb 3C11 in binding to mouse splenocytes.

The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 711-nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD1d
Applications Flow Cytometry
Classification Monoclonal
Clone 1B1
Concentration 0.2mg/mL
Conjugate Brilliant Violet 711
Description Cd1d1; Cd1.1; Cd1a; Cd1d; Ly-38
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse Cd1.1 cDNA-transfected RMA-S mouse T lymphoma and L929 cells
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.