Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD26 Rat anti-Mouse, Brilliant Violet 650, Clone: H194-112, BD Optibuild™

Catalog No. BDB740474 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740474 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740474 Supplier BD Biosciences Supplier No. 740474
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The H194-112 monoclonal antibody specifically binds to CD26, which is also known as, Thymocyte-activating molecule (THAM), or dipeptidyl peptidase IV (DPP IV, Dpp4). CD26 is a ∽220- kDa dimer formed of identical type-II transmembrane core polypeptides which undergo variable post-translational modifications. It is a multi-functional molecule with both ectopeptidase and signal-transducing activities. Studies with specific DPP IV inhibitors suggest that the enzymatic activity is involved in the mediation of T-cell activation events. The expression of CD26 is developmentally regulated in the thymus. Resting lymphoid cells of the bone marrow and peripheral B and T lymphocytes express low levels of CD26; bone-marrow and peritoneal myeloid cells do not. CD26 is also found on epithelial cells in the kidney, liver, small intestine, and lung. Cross-linked H194-112 mAb induces proliferation of immature and mature thymocytes in the presence of either IL-1 plus IL-2 or PMA; addition of IL-2 or IL-4 to PMA further enhances the activation.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 650-nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD26
Applications Flow Cytometry
Classification Monoclonal
Clone H194-112
Concentration 0.2mg/mL
Conjugate Brilliant Violet 650
Description Cd26; Dpp4; dipeptidylpeptidase 4; THAM, DPP IV
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Cell-surface molecules solubilized from BALB/c mouse thymocytes
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.