Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD276 Rat anti-Mouse, Brilliant Violet 605, Clone: MIH32, BD Optibuild™

Catalog No. BDB740435 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740435 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740435 Supplier BD Biosciences Supplier No. 740435
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The MIH32 monoclonal antibody specifically binds to CD276, also known as B7-H3 (B7 homolog 3). CD276 is a type I transmembrane glycoprotein and member of the B7-family of regulatory proteins. The expression of B7-H3 can be induced on T cells, natural killer (NK) cells and antigen presenting cells. B7-H3 is up-regulated during the differentiation of monocytes into dendritic cells or during the interaction between dendritic cells and regulatory T cells. In addition, B7-H3 is found to be expressed on fibroblasts, fibroblast-like synoviocytes and epithelial cells. CD276 (B7-H3) can function as a positive or a negative regulator of T responses.

This antibody is conjugated to BD Horizon™ BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max of 602-nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20-nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605-nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD276
Applications Flow Cytometry
Classification Monoclonal
Clone MIH32
Concentration 0.2mg/mL
Conjugate Brilliant Violet 605
Description Cd276; B7-H3; B7 homolog 3; B7h3; B7RP-2; Costimulatory molecule
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse B7-H3 Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.