Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD278 Rat anti-Mouse, Brilliant Violet 786, Clone: 7E.17G9, BD Optibuild™

Catalog No. BDB740955 Shop All BD Biosciences Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740955 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. BDB740955 missing translation for 'mfr' BD Biosciences missing translation for 'supplierNo' 740955
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 7E.17G9 monoclonal antibody specifically binds to CD278, the I nducible Cos timulatory molecule (ICOS), a 47-57 kDa homodimeric glycoprotein of the CD28 family of costimulatory molecules. ICOS is expressed on subpopulations of CD4-CD8- and CD4+CD8- (but not CD4-CD8+ or CD4+CD8+) thymocytes, on some T-cell lines, and on small numbers of peripheral leukocytes. It is upregulated on T lymphocytes following activation via the T-cell receptor. The T-cell activation marker H4 is the same molecule as ICOS. ICOS is a costimulatory receptor, and its ligand on antigen-presenting cells has been called B7RP-1, GL50, B7h, B7-H2, or LICOS. There is considerable evidence that the interaction of ICOS with its ligand is involved in the regulation of many, but not all, T-cell-mediated immune responses.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD278
Applications Flow Cytometry
Classification Monoclonal
Clone 7E.17G9
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Description ICOS; Ailim; CCLP; CRP-1; Ly115; Inducible T-cell costimulator
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse Icos cDNA and ICOS hexahistidine fusion protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.