Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD31 Rat anti-Mouse, Brilliant Violet 650, Clone: 390, BD Optibuild™

Catalog No. BDB740483 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740483 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740483 Supplier BD Biosciences Supplier No. 740483
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 390 monoclonal antibody specifically binds to CD31, also known as PECAM-1 (platelet endothelial cell adhesion molecule). CD31 is a ∽130 kDa integral membrane glycoprotein, a member of the immunoglobulin superfamily, that mediates homophilic and heterophilic cell-cell adhesion. CD31 is expressed constitutively on the surface of adult and embryonic endothelial cells and is weakly expressed on many peripheral leukocytes and platelets. It has also been detected on bone marrow-derived hematopoietic stem cells and embryonic stem cells. CD31 is involved in the transendothelial emigration of neutrophils, and neutrophil PECAM-1 appears to be down-regulated after extravasation into inflamed tissues. Multiple alternatively spliced isoforms are detected during early post-implantation embryonic development; this alternative splicing is involved in regulation of ligand specificity. CD38 and vitronectin receptor ( α v β 3 integrin, CD51/CD61) are proposed to be ligands for CD31. CD31-mediated endothelial cell-cell interactions are involved in angiogenesis. The 390 mAb inhibits a variety of in vitro and in vivo functions mediated by CD31.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 650-nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD31
Applications Flow Cytometry
Classification Monoclonal
Clone 390
Concentration 0.2mg/mL
Conjugate Brilliant Violet 650
Description Platelet endothelial cell adhesion molecule; Pecam1; PECAM-1; Pecam-1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen C3H/HeJ mouse hematopoietic progenitor cell line 32D
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.