Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD41 Rat anti-Mouse, Brilliant Violet 650, Clone: MWReg30, BD Optibuild™

Catalog No. BDB740504 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740504 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740504 Supplier BD Biosciences Supplier No. 740504
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The MWReg30 monoclonal antibody specifically binds to CD41, which is also known as glycoprotein IIb (gpIIb, GP IIb), Integrin α IIb chain, or Integrin alpha 2b. CD41 is a transmembrane glycoprotein that is encoded by Itga2b . CD41 associates with I ntegrin β3 chain (g pIIIa or CD61) to form the gpIIb/IIIa (CD41/CD61) complex. CD41/CD61 is expressed on platelets, megakaryocytes, and early hematopoietic progenitors. The integrin complex binds to fibrinogen, fibronectin, vitronectin, von Willebrand factor, and thrombospondin. It is important for platelet adhesion and aggregation, and it may play a role in osteolytic tumor metastasis.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD41
Applications Flow Cytometry
Classification Monoclonal
Clone MWReg30
Concentration 0.2mg/mL
Conjugate Brilliant Violet 650
Description Itga2b; Integrin alpha 2b; Integrin alpha-IIb; Integrin α IIb; GpIIb
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse Platelets
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.