Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45RA Rat anti-Mouse, Brilliant Violet 711, Clone: 14.8, BD Optibuild™

Catalog No. BDB740681 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740681 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740681 Supplier BD Biosciences Supplier No. 740681
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 14.8 monoclonal antibody specifically recognizes an exon A-dependent epitope of the CD45 protein, which is found at high density on B cells and at low density on peripheral T cytotoxic/suppressor cells and a very small subset of thymocytes. Nearly all B-lineage cells, including B-cell precursors in fetal liver and adult bone marrow and Ig-secreting cells, but not hematopoietic stem cells or myeloid progenitors, have been reported to be detectable by mAb 14.8. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family: Its intracellular (COOH-terminal) region contains two PTP catalytic domains, and the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated A, B, and C, respectively), plus, differing levels of glycosylation. The CD45 isoforms detected in the mouse are cell type-, maturation-, and activation state-specific. The CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction. mAb 14.8 has been reported to enhance the proliferative effect of PHA on purified spleen T cells, possibly by replacing a signal normally delivered by accessory cells, to enhance isotype switching during in vitro B-cell responses, and to inhibit antigen-induced p21 [ras] activation.

The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 711-nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD45RA
Applications Flow Cytometry
Classification Monoclonal
Clone 14.8
Concentration 0.2mg/mL
Conjugate Brilliant Violet 711
Description Ptprc; CD45R; CD45; LCA; Leukocyte common antigen; Ly-5; Lyt-4
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Radiation-induced NZC mouse B lymphoma WEHI-279
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.