Learn More
CD45RB Rat anti-Mouse, Brilliant Violet 650, Clone: 16A, BD Optibuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 740449
Description
The 16A monoclonal antibody specifically recognizes an exon B-dependent epitope of CD45 lycoprotein, which is found at high density on peripheral B cells, T cytotoxic/suppressor cells, subset of T helper cells, and most thymocytes and at low density on macrophages and dendritic cells.The level of CD45RB expression appears to decrease as T lymphocytes progress from naive to memory cells. In addition, subpopulations of CD4+ T cells which express high and low levels of D45RB have different cytokine secretion profiles and mediate distinct immunological functions. CD25+ D4+ regulatory T (Treg) lymphocytes which control intestinal inflammation and autoimmunity express low levels of D45RB. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family: Its intracellular (COOH-terminal) region contains two PTP catalytic domains, and the extracellular region is highly variable due to alternative splicing of exons (designated A, B, and C, respectively), plus differing levels of glycosylation. The CD45 isoforms detected in the mouse are cell type-, maturation-, and activation state-specific. The CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction.
The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 650-nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

Specifications
CD45RB | |
Monoclonal | |
0.2mg/mL | |
Ptprc; CD45R; CD45; LCA; Leukocyte common antigen; Ly-5; Lyt-4 | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
16A | |
Brilliant Violet 650 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Cloned Mouse TH2 cell lines | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only