Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD86 Rat anti-Mouse, Brilliant Violet 650, Clone: PO3, BD Optibuild™

Catalog No. BDB740501 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740501 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740501 Supplier BD Biosciences Supplier No. 740501
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The PO3 monoclonal antibody specifically binds to the B7-2 (CD86) costimulatory molecule, which is expressed on a broad spectrum of leukocytes, including B lymphocytes, T lymphocytes, macrophages, dendritic cells, and Langerhans cells, plus astrocytes. CD86 is expressed at low levels by freshly explanted peripheral B and T cells, and its expression is substantially increased by a variety of T and B cell-specific stimuli with a peak expression after 18-42 hours of culture. In contrast to most naive CD4+ T cells, memory CD4+ T cells express B7-2, both at the mRNA and protein levels. CD86, a ligand for CD28 and CTLA-4, is one of the accessory molecules that plays an important role in T cell-B cell costimulatory interactions. It has been shown to be involved in immunoglobulin class switching and triggering of mouse NK cell-mediated cytotoxicity. CD80 (B7-1) is an alternate ligand for CD28 and CTLA-4. PO3 antibody blocks the in vitro stimulation of T-cell proliferation of T-cell proliferation by soluble anti-CD3e antibody (mAb 145-2C11, Cat. No. 553057) in the presence of CD86-expressing accessory cells. In vivo administration of PO3 mAb can inhibit much of the autoantibody production in (NZB x NZW) F1 mice; and in combination with an anti-CD80 mAb, it can prevent the development and progression of mouse systemic lupus erythematosus-like autoimmune disease.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 650-nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD86
Applications Flow Cytometry
Classification Monoclonal
Clone PO3
Concentration 0.2mg/mL
Conjugate Brilliant Violet 650
Description Cd86; B7-2; B7.2; Ly58; Ly-58; ETC-1; MB7-2; TS/A2; Cd28l2
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen BALB/c Mouse B Leukemia Cell Line BCL1
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.