Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD94 Rat anti-Mouse, Brilliant Violet 786, Clone: 18d3, BD Optibuild™

Catalog No. BDB740951 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740951 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740951 Supplier BD Biosciences Supplier No. 740951
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 18d3 monoclonal antibody specifically recognizes CD94 on all NK cells, NK1.1- or DX5-positive T lymphocytes (NK-T cells), and a subset of CD8-positive T lymphocytes in most strains tested (eg, A/J, AKR/J, BALB/c, C3H/He, C57BL/6, CBA/J, DBA/1, FVB/N, 129/Sv, NOD, SWR, and most DBA/2 substrains, but not DBA/2J). DBA/2J mice do not express CD94.3 CD94 is also expressed on CD8+ T lymphocytes activated in vivo. CD94 is a type-II transmembrane protein with an extracellular lectin-like domain and a short cytoplasmic tail which is not believed to have any signalling function. Heterodimers of CD94 with NKG2A, NKG2C, or NKG2E recognize Qa-1 (a non-classical MHC class I antigen) presenting the Qdm peptide. Studies on CD94/NKG2 heterodimers on human NK cells have demonstrated that the NKG2 components mediate signal transduction for the receptor, NKG2A being inhibitory and NKG2C being stimulatory. Similarly, the mouse NKG2A molecule contains two intracytoplasmic sequences which resemble the ITIM ( Immunoreceptor Tyrosine-based Inhibitory Motif ) consensus sequence. CD94/NKG2 receptors appear on fetal NK cells before the Ly- 49 MHC class I receptors, suggesting that CD94/NKG2 receptors and their ligand, Qa-1, may play a role in maintenance of self-tolerance in developing NK cells.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 786-nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60-nm filter).

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD94
Applications Flow Cytometry
Classification Monoclonal
Clone 18d3
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Description Cd94; Klrd1; Killer cell lectin-like receptor, subfamily D, member 1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Transfected cell line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.