Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Early B Lineage Rat anti-Mouse, Brilliant Violet 421, Clone: 493, BD Optibuild™

Catalog No. BDB740064 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740064 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740064 Supplier BD Biosciences Supplier No. 740064
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 493 antibody reacts with a cell-surface protein of 130-140 kDa expressed on immature B lymphocytes and a small fraction of newly formed B cells, but not on mature B lymphocytes. The antigen’s distribution was defined through the use of antibodies to CD24 (Heat Stable Antigen), IgM, IgD, and CD45R/B220, which are commonly used to discriminate B-cell maturation stages. 493 mAb reacts with the majority of B220+ cells in bone marrow and a fraction of B220+ B cells in spleen, which are CD24[high], IgM[high], and IgD[low]. Cells binding 493 mAb were not detected in thymus, lymph nodes, or peritoneal cavity. This result suggested that 493 mAb does not stain B-1 B cells (CD5+ B lymphocytes), which are particularly found in the peritoneal cavity. The 493 mAb does not seem to have any biological effect when incubated with immature B lymphocytes. It has been observed that the staining pattern of mAb 493 is similar to that of mAb AA4.1, that both antibodies precipitate molecules of the same molecular weight, and that staining by mAb AA4.1 is not blocked by mAb 493, suggesting that the antibodies recognize separate epitopes of the same Early B Lineage antigen.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Early B Lineage
Applications Flow Cytometry
Classification Monoclonal
Clone 493
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Description Cd93; Ly68; C1qr1; C1qrp; Complement component C1q receptor
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Pro-B cell line R2BFL derived from the fetal liver of a RAG-2-/-Bcl2-transgenic mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.