Learn More
Early B Lineage Rat anti-Mouse, Brilliant Violet 650, Clone: 493, BD Optibuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 740548
Description
The 493 antibody reacts with a cell-surface protein of 130-140 kDa expressed on immature B lymphocytes and a small fraction of newly formed B cells, but not on mature B lymphocytes. The antigen’s distribution was defined through the use of antibodies to CD24 (Heat Stable Antigen), IgM, IgD, and CD45R/B220, which are commonly used to discriminate B-cell maturation stages. 493 mAb reacts with the majority of B220+ cells in bone marrow and a fraction of B220+ B cells in spleen, which are CD24[high], IgM[high], and IgD[low]. Cells binding 493 mAb were not detected in thymus, lymph nodes, or peritoneal cavity. This result suggested that 493 mAb does not stain B-1 B cells (CD5+ B lymphocytes), which are particularly found in the peritoneal cavity. The 493 mAb does not seem to have any biological effect when incubated with immature B lymphocytes. It has been observed that the staining pattern of mAb 493 is similar to that of mAb AA4.1, that both antibodies precipitate molecules of the same molecular weight, and that staining by mAb AA4.1 is not blocked by mAb 493, suggesting that the antibodies recognize separate epitopes of the same Early B Lineage antigen.
The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 650-nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

Specifications
Early B Lineage | |
Monoclonal | |
0.2mg/mL | |
Cd93; Ly68; C1qr1; C1qrp; Complement component C1q receptor | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
493 | |
Brilliant Violet 650 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Pro-B cell line R2BFL derived from the fetal liver of a RAG-2-/-Bcl2-transgenic mouse | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only